missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Cerium(Iv) Nitrate, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 100 g |
Linear Formula | Ce(NH4)2(NO3)6 |
UN Number | 3085 |
Formula Weight | 548.22g/mol |
Percent Purity | ≥98.5% (RT) |
Grade | ACS/puriss. p.a. |
Packaging | Plastic Bottle |
Chemical Name or Material | Ammonium Cerium(Iv) Nitrate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction