missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Cerium(Iv) Nitrate, Honeywell Fluka™

Product Code. 15632190
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss. p.a., ACS Reagent,≥98.5% (RT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16774-21-3
MDL Number MFCD00151121

Specifications

Quantity 500 g
Linear Formula Ce(NH4)2(NO3)6
UN Number 3085
Formula Weight 548.22g/mol
Percent Purity ≥98.5% (RT)
Grade ACS/puriss. p.a.
Packaging Plastic Bottle
Chemical Name or Material Ammonium Cerium(Iv) Nitrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.