missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium cerium(IV) sulfate dihydrate, 99+%, extra pure, Thermo Scientific Chemicals
£26.03 - £338.69
Chemical Identifiers
CAS | 10378-47-9 |
---|---|
Molecular Formula | H16CeN4O16S4·2H2O |
Molecular Weight (g/mol) | 632.54 |
MDL Number | MFCD00149417 |
Synonym | Ceric ammonium sulfate |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10194780
|
Thermo Scientific Acros
206310050 |
5 g | Glass bottle |
£26.03
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10398480
|
Thermo Scientific Acros
206311000 |
100 g | Glass bottle |
£92.26
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10219203
|
Thermo Scientific Acros
206315000 |
500 g | Glass bottle |
£338.69
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10378-47-9 | |
632.54 | |
Ceric ammonium sulfate |
H16CeN4O16S4·2H2O | |
MFCD00149417 |
Specifications
130.0°C | |
Orange | |
99+% | |
(NH4)4Ce(SO4)4·2H2O | |
08,80 | |
Solubility in water: hydrolysis. Other solubilities: insoluble in ethanol | |
632.54 | |
Extra Pure |
10378-47-9 | |
Crystalline Powder | |
H16CeN4O16S4·2H2O | |
MFCD00149417 | |
Ceric ammonium sulfate | |
632.54 | |
≥99% | |
Ammonium cerium(IV) sulfate dihydrate, Extra Pure |
Safety and Handling
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Ammonium cerium(IV) sulfate dihydrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ammonium cerium(IV) sulfate dihydrate, 99+%, extra pure, Thermo Scientific Chemicals