missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Chloride, 99+%, Extra Pure, SLR, Fisher Chemical™

Product Code. 10488010 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 g
2 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
2kg
500g
5kg
This item is not returnable. View return policy

Product Code. 10488010

Brand: Fisher Chemical A/3880/61

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
IUPAC Name amine hydrochloride
SMILES N.Cl

Specifications

Melting Point 340°C
Color White
pH 5
Physical Form Solid
Quantity 2 kg
Formula Weight 53.49g/mol
Packaging Plastic powder jar
Vapor Pressure 1.3mbar 160
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.