Learn More
Ammonium chloride, 99+%, pure, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 340.0°C |
Color | White |
Physical Form | Crystalline Powder and/or Chunks |
Quantity | 1 kg |
Assay Percent Range | 99% min. (Argentometry) |
Linear Formula | NH3·HCl |
Fieser | 15,13 |
Merck Index | 15, 506 |
Solubility Information | Solubility in water: 370g/L (20°C). Other solubilities: soluble in glycerol,slightly soluble in alcohol,practically insoluble in acetone,ether and,ethyl acetate |
IUPAC Name | azanium;chloride |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0466752 | Certificate of Analysis | 11/10/2024 | 123340050,123340250,123340251,123340010 |
A0466751 | Certificate of Analysis | 11/10/2024 | 123340050,123340250,123340251,123340010 |
A0461804 | Certificate of Analysis | 12/04/2024 | 123340050,123340250,123340251,123340010 |
A0461805 | Certificate of Analysis | 12/04/2024 | 123340050,123340250,123340251,123340010 |
A0456560 | Certificate of Analysis | 29/09/2023 | 123340050,123340250,123340251,123340010 |
Safety and Handling

- Ammonium chloride
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.