missing translation for 'onlineSavingsMsg'
Learn More
Ammonium chloride, 99+%, pure, Thermo Scientific Chemicals Product Code.: 10276873

Ammonium chloride, 99+%, pure, Thermo Scientific Chemicals

Product Code. 10276873
25 kg, Fibre drum
Click to view available options
Quantity:
25 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic Bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10276873

Brand: Thermo Scientific Acros 123340251

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ammonium chloride, 99+%, H4ClN, CAS Number-12125-02-9, amchlor, ammoneric, ammoniumchlorid, ammonium chloride, ammonium chloride nh4 cl, ammoniumklorid, ammonium muriate, darammon, sal ammoniac, salmiac, 1kg, 99% min. (Argentometry), CHEBI:31206, White, 1.5200g/mL, 235-186-4, 15,13, 53.49, Pure

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
PubChem CID 25517
ChEBI CHEBI:31206
SMILES N.Cl

Specifications

Melting Point 340.0°C
Color White
Physical Form Crystalline Powder and/or Chunks
Quantity 25 kg
Assay Percent Range 99% min. (Argentometry)
Linear Formula NH3·HCl
Fieser 15,13
Merck Index 15, 506
Solubility Information Solubility in water: 370g/L (20°C). Other solubilities: soluble in glycerol,slightly soluble in alcohol,practically insoluble in acetone,ether and,ethyl acetate
IUPAC Name amine hydrochloride
Formula Weight 53.49
Percent Purity ≥99%
Grade Pure
Packaging Fibre drum
Density 1.5200g/mL
Chemical Name or Material Ammonium chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466752 Certificate TypeCertificate of Analysis Date11/10/2024 Catalog Number123340050,123340250,123340251,123340010
Lot NumberA0466751 Certificate TypeCertificate of Analysis Date11/10/2024 Catalog Number123340050,123340250,123340251,123340010
Lot NumberA0461804 Certificate TypeCertificate of Analysis Date12/04/2024 Catalog Number123340050,123340250,123340251,123340010
Lot NumberA0461805 Certificate TypeCertificate of Analysis Date12/04/2024 Catalog Number123340050,123340250,123340251,123340010
Lot NumberA0456560 Certificate TypeCertificate of Analysis Date29/09/2023 Catalog Number123340050,123340250,123340251,123340010
compliance-icons
Product Identifier
  • Ammonium chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium chloride, 99+%, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.