missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium chloride, for analysis, Thermo Scientific Chemicals
16.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Especificaciones
Melting Point | 340.0°C |
Color | White |
Physical Form | Crystalline Powder and/or Crystals |
Quantity | 250 g |
Linear Formula | NH3·HCl |
Merck Index | 15, 506 |
Solubility Information | Solubility in water: 370g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol, practically insoluble in acetone, ether and, ethyl acetate |
IUPAC Name | amine hydrochloride |
Formula Weight | 53.49 |
Percent Purity | ≥99.0% |
Mostrar más |
Seguridad y manipulación
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Nombre del producto
- Ammonium chloride
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'