missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Chloride, Certified AR, for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

Product Code. 10151750
Click to view available options
Quantity:
500 g
2 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar

H4ClN, CAS Number-12125-02-9, 5kg, White, 53.49g/mol, 340 deg.C, 11420, Plastic keg, Solid, 1.3mbar 160

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
IUPAC Name amine hydrochloride
SMILES N.Cl

Specifications

Melting Point 340°C
Color White
pH 5
Physical Form Solid
Quantity 5 kg
Formula Weight 53.49g/mol
Packaging Plastic keg
Vapor Pressure 1.3mbar 160
compliance-icons
Product Identifier
  • Ammonium chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'