missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Chloride, Certified AR, for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
H4ClN, CAS Number-12125-02-9, 5kg, White, 53.49g/mol, 340 deg.C, 11420, Plastic keg, Solid, 1.3mbar 160
£66.20 - £499.00
Chemical Identifiers
CAS | 12125-02-9 |
---|---|
Molecular Formula | ClH4N |
Molecular Weight (g/mol) | 53.49 |
MDL Number | MFCD00011420 |
InChI Key | NLXLAEXVIDQMFP-UHFFFAOYSA-N |
IUPAC Name | amine hydrochloride |
SMILES | N.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10060030
|
Fisher Chemical
A/3920/53 |
500 g | Plastic powder jar | ||||||
10213900
|
Fisher Chemical
A/3920/61 |
2 kg | Plastic powder jar |
£212.00
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10151750
|
Fisher Chemical
A/3920/65 |
5 kg | Plastic keg |
£499.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
H4ClN, CAS Number-12125-02-9, 5kg, White, 53.49g/mol, 340 deg.C, 11420, Plastic keg, Solid, 1.3mbar 160
Chemical Identifiers
12125-02-9 | |
53.49 | |
NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
N.Cl |
ClH4N | |
MFCD00011420 | |
amine hydrochloride |
Specifications
340°C | |
White | |
Solid | |
MFCD00011420 | |
N.Cl | |
53.49 | |
1.3mbar 160 |
12125-02-9 | |
5 | |
ClH4N | |
NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
amine hydrochloride | |
53.49g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Ammonium chloride
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.