missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Chloride, Reagent Grade, ≥99.5%, Honeywell™
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly Bottle
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Melting Point | 338°C |
Color | Colorless |
pH | 4.6 to 6 (at 20°C) |
Physical Form | Solid |
Quantity | 500 g |
Linear Formula | NH4Cl |
Beilstein | 4371014 |
SMILES | N.Cl |
Formula Weight | 53.49g/mol |
Percent Purity | ≥99.5% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'