Learn More
STEMCELL Technologies Ammonium Chloride Solution
Reagent for lysis of red blood cells
Brand:  STEMCELL Technologies 07850
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 15570226
  • £172.80 / 500mL
Estimated Shipment: 24-07-2024
to see stock.
Add to basket
Description
Description
  • Ammonium Chloride Solution is recommended for the lysis of red blood cells (RBCs) in preparations of human and mouse peripheral blood, spleen, or bone marrow cells
  • It is buffered and optimized for gentle lysis of erythrocytes, with minimal effect on leukocytes
  • It does not contain a fixative agent, therefore leukocytes are viable following RBC lysis
Specifications
Specifications
500 mL
Lysis of Red Blood Cells