missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Citrate Dibasic, Honeywell Fluka™

Product Code. 15660910
Click to view available options
Quantity:
100 g
500 g

Chemical Identifiers

CAS 3012-65-5
Molecular Formula C6H14N2O7
Molecular Weight (g/mol) 226.19
MDL Number MFCD00013068
InChI Key YXVFQADLFFNVDS-UHFFFAOYSA-N
Synonym ammonium citrate dibasic, ammonium hydrogencitrate, diammonium hydrogen citrate, citric acid diammonium salt, citric acid ammonium salt, di-ammonium hydrogen citrate, citro diamine, citric acid diamine, ammonium citrate, dibasic, acs, ammonium citrate dibasic, puriss
PubChem CID 13710713
ChEBI CHEBI:63076
SMILES [NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O

Specifications

Quantity 500 g
Linear Formula HOC(CO2H)(CH2CO2NH4)2
Beilstein 4925760
IUPAC Name azane;2-hydroxypropane-1,2,3-tricarboxylic acid
Formula Weight 226.18g/mol
Percent Purity ≥99% (T)
Grade ACS/puriss. p.a.
Residue after Ignition ≤0.01% (as SO4)
Density Vapor Density: 1.8 (vs air)
Chemical Name or Material Ammonium Citrate Dibasic
compliance-icons
Product Identifier
  • Ammonium citrate dibasic
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Ammonium citrate dibasic
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'