Learn More
Ammonium cobalt(II) sulfate hexahydrate, 98%, Thermo Scientific Chemicals
£29.50 - £293.00
Chemical Identifiers
CAS | 13586-38-4 |
---|---|
Molecular Formula | (NH4)2Co(SO4)2·6H2O |
MDL Number | MFCD00151049 |
Synonym | Cobalt(II) ammonium sulfate; Cobaltous ammonium sulfate |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11403344
|
Thermo Scientific Alfa Aesar
A10627.14 |
25 g |
£29.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11413344
|
Thermo Scientific Alfa Aesar
A10627.22 |
100 g |
£81.90
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11423344
|
Thermo Scientific Alfa Aesar
A10627.36 |
500 g |
£293.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13586-38-4 | |
(NH4)2Co(SO4)2·6H2O | |
MFCD00151049 | |
Cobalt(II) ammonium sulfate; Cobaltous ammonium sulfate |
Specifications
∼120°C (decomposition) | |
(NH4)2Co(SO4)2·6H2O | |
UN3077 | |
395.21 (287.13 Anhydrous) | |
Odorless | |
1.902 |
13586-38-4 | |
MFCD00151049 | |
Cobalt(II) ammonium sulfate; Cobaltous ammonium sulfate | |
98% | |
Hygroscopic | |
Ammonium cobalt (II) sulfate hexahydrate |
Safety and Handling
GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.
P201-P202-P261-P264b-P270-P272-P280g-P281-P301+P312-P302+P352-P308+P313-P330-P333+P313-P363-P501c
H302-H317-H350
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'einecsNumber' : 237-043-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only