missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium copper(II) chloride dihydrate, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Ammonium copper(II) chloride dihydrate is used in the determination iron of content in steel. It is also used in coatings, printing ink, rubber, glass, leather and cosmetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 110°C (decomposition) |
Color | Blue |
Physical Form | Crystals |
Quantity | 100 g |
Merck Index | 14,514 |
Solubility Information | Soluble in water,acids and alcohol; Slightly soluble in ammonia. |
Formula Weight | 277.46 (241.43 Anhydrous) |
Density | 1.993 g/mL |
Chemical Name or Material | Ammonium copper(II) chloride dihydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'