missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ Ammonium Cuvette Tests

£137.00 - £140.00

Specifications

Certifications/Compliance ISO 7150-1, DIN 38406 E5-1
No. of Tests 25
Storage Requirements 2°C to 8°C
Test Method Indophenol Blue
Type Ammonium Cuvette Test
View More Specs

Products 2
Product Code Brand Range Price Quantity & Availability  
Product Code Brand Range Price Quantity & Availability  
10566844
View Documents
Hach Lange™
LCK302
47 to 130mg/L NH4-N
£140.00
Each
Please to purchase this item. Need a web account? Register with us today!
10165551
View Documents
Hach Lange™
LCK305
0 to 12mg/L NH4-N
£137.00
Pack of 25
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

ISO 7150-1, DIN 38406 E5-1
2°C to 8°C
Ammonium Cuvette Test
25
Indophenol Blue
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • LatoN, Sample cuvette

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 3
  • Flammable liquid Category 3

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H360FD-May damage fertility. May damage the unborn child.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Sulphuric acid, Phosphoric acid, Sodiumhydroxide, Potassiumnitrate, dipotassium peroxodisulphate,

SDS
Documents

Documents

Product Certifications