missing translation for 'onlineSavingsMsg'
Learn More

Ammonium dichromate, 99%, Thermo Scientific Chemicals

Product Code. 10752295
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-09-5
Molecular Formula H8Cr2N2O7
Molecular Weight (g/mol) 252.07
MDL Number MFCD00010879
InChI Key JOSWYUNQBRPBDN-UHFFFAOYSA-P
Synonym ammonium dichromate, ammonium bichromate, diammonium dichromate, ammonium dichromate vi, ammoniumbichromaat dutch, ammoniumdichromaat dutch, ammoniumdichromat german, unii-5j18bp595g, hsdb 481
PubChem CID 24600
IUPAC Name diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]

Specifications

Melting Point 170.0°C
Color Orange
Physical Form Adhering Crystals
Quantity 100 g
Assay Percent Range 98.5% min. (Iodometry)
Linear Formula (NH4)2Cr2O7
Merck Index 15, 512
Solubility Information Solubility in water: 360g/L (20°C). Other solubilities: soluble in alcohol
Formula Weight 252.07
Percent Purity 99%
Loss on Drying 3% max. (105°C, 4 hrs)
Packaging Plastic bottle
Chemical Name or Material Ammonium dichromate, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'