missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Dihydrogen Orthophosphate, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
Description
Ammonium Dihydrogen Orthophosphate, Certified AR for Analysis
Specifications
Specifications
Melting Point | 190°C |
Color | White |
pH | 3.8 to 4.4 |
Physical Form | Solid |
Quantity | 500 g |
Formula Weight | 115.03g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'