missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium dihydrogen phosphate, 99.999%, (trace metal basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
Specifications
Specifications
Melting Point | 190.0°C |
Color | White or Colorless |
Physical Form | Crystals |
Quantity | 25 g |
Assay Percent Range | 99.999% |
Linear Formula | NH4H2PO4 |
Merck Index | 15,541 |
Solubility Information | Solubility in water: 368g/L (20°C). Other solubilities: slightly soluble in alcohol, practically insoluble in acetone |
Formula Weight | 115.03 |
Percent Purity | 99.999% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'