missing translation for 'onlineSavingsMsg'
Learn More

Ammonium dihydrogen phosphate, 99+%, for analysis, Thermo Scientific Chemicals

Product Code. 15199213
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic Drum
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-76-1
Molecular Formula H6NO4P
Molecular Weight (g/mol) 115.03
MDL Number MFCD00003396
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
Synonym ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
PubChem CID 24402
ChEBI CHEBI:62982
IUPAC Name phosphoric acid amine
SMILES N.OP(O)(O)=O

Specifications

Melting Point 190.0°C
Color White
pH 4 to 4.5 (5% soln. at 20°C)
Physical Form Crystalline Solid
Quantity 10 kg
Assay Percent Range 99+%
Linear Formula NH4H2PO4
Merck Index 15,541
Solubility Information Solubility in water: 368g/L (20°C). Other solubilities: slightly soluble in alcohol, practically insoluble in acetone
Formula Weight 115.03
Percent Purity ≥99%
Grade Analytical
Packaging Plastic Drum
Density 1.8g/mL
Chemical Name or Material Ammonium Dihydrogen Phosphate
Show More Show Less
Product Identifier
  • Ammonium dihydrogen phosphate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'