Learn More
Ammonium dihydrogen phosphate, ACS, 98.0% min, Thermo Scientific Chemicals
£41.50 - £108.00
Chemical Identifiers
CAS | 7722-76-1 |
---|---|
Molecular Formula | H6NO4P |
Molecular Weight (g/mol) | 115.03 |
MDL Number | MFCD00003396 |
InChI Key | LFVGISIMTYGQHF-UHFFFAOYSA-N |
Synonym | ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate |
PubChem CID | 24402 |
ChEBI | CHEBI:62982 |
IUPAC Name | phosphoric acid amine |
SMILES | N.OP(O)(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11307347
|
Thermo Scientific Alfa Aesar
011598.36 |
500 g |
£41.50
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11317347
|
Thermo Scientific Alfa Aesar
011598.A3 |
2 kg |
£108.00
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Preparation of baking powder with sodium bicarbonate, in fermentation.Ammonium dihydrogen phosphate is widely utilized as a fertilizer, which provides phosphorous and nitrogen for plants. It is used as a dry fire extinguisher. It is a well known nonlinear optical material for various optoelectronic applications. It is also used in an emerald green, amethyst and aquamarine crystal growing box kit for children.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7722-76-1 | |
115.03 | |
LFVGISIMTYGQHF-UHFFFAOYSA-N | |
24402 | |
phosphoric acid amine |
H6NO4P | |
MFCD00003396 | |
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate | |
CHEBI:62982 | |
N.OP(O)(O)=O |
Specifications
190°C | |
Crystalline | |
H6NO4P | |
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate | |
LFVGISIMTYGQHF-UHFFFAOYSA-N | |
phosphoric acid amine | |
24402 | |
115.03 | |
Odorless | |
Ammonium dihydrogen phosphate |
7722-76-1 | |
98% min. | |
MFCD00003396 | |
Freely soluble in water. Slightly soluble in alcohol. Insoluble in acetone | |
N.OP(O)(O)=O | |
115.03 | |
CHEBI:62982 | |
ACS | |
1.803 g/cm3 |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste.
H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
missing translation for 'einecsNumber' : 231-764-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only