missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium fluoride, Puriss. p.a., ACS Reagent, ≥98%, Honeywell Fluka™
Click to view available options
Quantity:
100 g
1 kg
50 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS Max % | 100.0000% |
Quantity | 50 kg |
Linear Formula | (CH3OCH2CH2)2O |
MDL Number | MFCD00008503 |
UN Number | UN2505 |
Formula Weight | 37.0 |
Percent Purity | >98% |
Grade | ACS Reagent |
Packaging | Fiberboard Box |
Residue after Ignition | ≤0.005% (as SO4) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'