missing translation for 'onlineSavingsMsg'
Learn More

Ammonium fluoride solution, Honeywell™

Semiconductor grade VLSI (Honeywell 17600), 40%

Brand:  Honeywell Research Chemicals 40292H-5L

Additional Details : Weight : 5.55000kg Transport : UN number : 3287 Chem class : 6.1 Pack group : III


Product Code. 16676173

  • £121.53 / 5 L
Estimated Shipment: 26-11-2024
to see stock.

Description

Description

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications

Specifications

12125-01-8,7732-18-5
≥50.00%
Semiconductor grade reagent
Plastic Can
Liquid
> 100°C (1.013 hPa)
<70.00%
37.04
5 L
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ammonium fluoride solution

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : ammonium fluoride

SDS
Documents

Documents

Certificates
Special Offers

Special Offers