Learn More
Ammonium Formate in Methanol, 10mM, with 0.05% Formic Acid, LC-MS, Thermo Scientific™
Mobile phase blend suitable for analysis such as vitamin D and drugs of abuse testing, and packaged in a borosilicate glass bottle that can be used directly on the LC-MS instrument.
Brand: Fisher Chemical MB122-1
83.50 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Ammonium Formate in Methanol, 10mM, with 0.05% Formic Acid, 0.57 to 0.69g/L (Ammonium Formate, w/v), CAS Number-540-69-2, 67-56-1, 64-18-6
- Suitable for use in LC/MS and UHPLC applications
- Pre-mixed for ease-of-use
- Mobile phase blend suitable for vitamin D and drugs of abuse testing
- Packaged in borosilicate glass bottles to prevent metal adduct formation, which can be used directly on the instrument
- Ammonium formate- a commonly used HPLC buffer that is compatible with LC/MS

Specifications
64.7°C | |
Mobile-phase solvent blend | |
3.5 (25°C) | |
1 L | |
MFCD00004595 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
UHPLC-MS | |
Borosilicate Glass Bottle | |
0.791 g/cm3 |
Ammonium Formate in Methanol, 10mM, with 0.05% Formic Acid | |
540-69-2, 67-56-1, 64-18-6 | |
Liquid | |
CH4O | |
ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny | |
CO | |
32.04 | |
10mM Ammonium Formate | |
128 at 20°C |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- 10mM Ammonium Formate in Methanol + 0.05% Formic Acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
- Flammable liquid Category 2
Hazard Statement
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
Supplemental information
- MIXTURE LIST-Contains : Methyl alcohol
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only