Learn More
Ammonium formate, 99%, Thermo Scientific Chemicals
£20.20 - £213.00
Chemical Identifiers
CAS | 540-69-2 |
---|---|
Molecular Formula | CH5NO2 |
Molecular Weight (g/mol) | 63.06 |
InChI Key | VZTDIZULWFCMLS-UHFFFAOYSA-N |
Synonym | ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny |
PubChem CID | 2723923 |
ChEBI | CHEBI:63050 |
IUPAC Name | azanium;formate |
SMILES | C(=O)[O-].[NH4+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10259690
|
Thermo Scientific Acros
401152500 |
250 g | Glass bottle |
£20.20
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10299640
|
Thermo Scientific Acros
401150010 |
1 kg | Plastic bottle |
£51.70
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11924891
|
Thermo Scientific Acros
401150050 |
5 kg | Plastic drum |
£213.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
540-69-2 | |
63.06 | |
ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny | |
CHEBI:63050 | |
C(=O)[O-].[NH4+] |
CH5NO2 | |
VZTDIZULWFCMLS-UHFFFAOYSA-N | |
2723923 | |
azanium;formate |
Specifications
115.0°C to 120.0°C | |
White | |
99% | |
HCO2NH4 | |
01,38; 13,230; 15,13 | |
ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny | |
VZTDIZULWFCMLS-UHFFFAOYSA-N | |
azanium;formate | |
2723923 | |
63.06 | |
Authentic |
540-69-2 | |
Crystalline Powder and/or Crystals | |
CH5NO2 | |
02,8 | |
15,52 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol | |
C(=O)[O-].[NH4+] | |
63.06 | |
CHEBI:63050 | |
99% | |
Ammonium formate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protect
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 208-753-9
- Ammonium formate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only