missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Formate, for HPLC, Honeywell Fluka™

for HPLC,≥99.0%

£71.39 - £280.35

Chemical Identifiers

CAS 540-69-2
Molecular Formula CH5NO2
Molecular Weight (g/mol) 63.056
MDL Number MFCD00013103
InChI Key VZTDIZULWFCMLS-UHFFFAOYSA-N
Synonym ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny
PubChem CID 2723923
ChEBI CHEBI:63050
IUPAC Name azanium;formate
SMILES C(=O)[O-].[NH4+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15681580
View Documents Promotion Details
Honeywell-Fluka
17843-50G
50 g
£71.39
50g
Please to purchase this item. Need a web account? Register with us today!
15671580
View Documents
Honeywell-Fluka
17843-250G
250 g
£280.35
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

540-69-2
63.056
VZTDIZULWFCMLS-UHFFFAOYSA-N
2723923
azanium;formate
CH5NO2
MFCD00013103
ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny
CHEBI:63050
C(=O)[O-].[NH4+]

Specifications

540-69-2
HCO2NH4
NONH for all modes of transport
ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny
VZTDIZULWFCMLS-UHFFFAOYSA-N
azanium;formate
2723923
63.06g/mol
1.26g/mL at 25°C
CH5NO2
MFCD00013103
3625095
Soluble in water 0.1g/mL at 25°C
C(=O)[O-].[NH4+]
63.056
CHEBI:63050
≥99%
Ammonium Formate
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 208-753-9

missing translation for 'rtecsNumber' : BQ6650000

Product Identifier
  • Ammonium formate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : ammonium formate

SDS
Documents

Documents

Product Certifications