missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Formate, Reagent grade, 97%, Honeywell Burdick & Jackson™

Product Code. 15651500
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, 97%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-69-2
Molecular Formula CH5NO2
Molecular Weight (g/mol) 63.056
MDL Number MFCD00013103
InChI Key VZTDIZULWFCMLS-UHFFFAOYSA-N
Synonym ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny
PubChem CID 2723923
ChEBI CHEBI:63050
IUPAC Name azanium;formate
SMILES C(=O)[O-].[NH4+]

Specifications

Melting Point 116°C
Color White
pH 6.0 to 7.0 (at 20°C)
Quantity 1 kg
Linear Formula HCO2NH4
UN Number NONH for all modes of transport
Beilstein 3625095
Formula Weight 63.06g/mol
Percent Purity 97%
Grade Reagent
Packaging Poly Bottle
Appearance Crystals/Granules
Density 1.26g/mL at 25°C
Chemical Name or Material Ammonium Formate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.