missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Formate, Reagent grade, 97%, Honeywell Burdick & Jackson™

Product Code. 15661500
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, 97%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-69-2
Molecular Formula CH5NO2
Molecular Weight (g/mol) 63.056
MDL Number MFCD00013103
InChI Key VZTDIZULWFCMLS-UHFFFAOYSA-N
Synonym ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny
PubChem CID 2723923
ChEBI CHEBI:63050
IUPAC Name azanium;formate
SMILES C(=O)[O-].[NH4+]

Specifications

Melting Point 116°C
Color White
pH 6.0 to 7.0 (at 20°C)
Quantity 2.5 kg
Linear Formula HCO2NH4
UN Number NONH for all modes of transport
Beilstein 3625095
Formula Weight 63.06g/mol
Percent Purity 97%
Grade Reagent
Packaging Poly Bottle
Appearance Crystals/Granules
Density 1.26g/mL at 25°C
Chemical Name or Material Ammonium Formate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'