Learn More
Ammonium hexachloroosmate(IV), 99.9% (metals basis), Os 42.5% min, Thermo Scientific Chemicals
£131.00 - £572.00
Chemical Identifiers
CAS | 12125-08-5 |
---|---|
Molecular Formula | Cl6H8N2Os |
Molecular Weight (g/mol) | 439.00 |
MDL Number | MFCD00010883 |
InChI Key | JRHMYOOMZKAPKT-UHFFFAOYSA-J |
Synonym | ammonium hexachloroosmate iv, nh4 2oscl6, osmium iv-ammonium chloride, diazanium;hexachloroosmium 2-, diammonium hexachloroosmiumdiuide, ammonium hexachloroosmiate iv , premion |
PubChem CID | 11729867 |
IUPAC Name | diazanium;hexachloroosmium(2-) |
SMILES | [NH4+].[NH4+].Cl[Os+4](Cl)(Cl)(Cl)(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11330118
|
Thermo Scientific Alfa Aesar
042874.03 |
1 g |
£131.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11320118
|
Thermo Scientific Alfa Aesar
042874.06 |
5 g |
£572.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ammonium hexachloroosmate(IV) acts as a pharmaceutical, organic and chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12125-08-5 | |
439.00 | |
JRHMYOOMZKAPKT-UHFFFAOYSA-J | |
11729867 | |
[NH4+].[NH4+].Cl[Os+4](Cl)(Cl)(Cl)(Cl)Cl |
Cl6H8N2Os | |
MFCD00010883 | |
ammonium hexachloroosmate iv, nh4 2oscl6, osmium iv-ammonium chloride, diazanium;hexachloroosmium 2-, diammonium hexachloroosmiumdiuide, ammonium hexachloroosmiate iv , premion | |
diazanium;hexachloroosmium(2-) |
Specifications
170° (sublimation) | |
Brown to Black | |
99.9% (Os 42.5% min.) (Metals basis) | |
MFCD00010883 | |
Soluble in water. | |
[NH4+].[NH4+].Cl[Os+4](Cl)(Cl)(Cl)(Cl)Cl | |
439.00 | |
438.99 | |
2.9 |
12125-08-5 | |
Crystalline | |
Cl6H8N2Os | |
ammonium hexachloroosmate iv, nh4 2oscl6, osmium iv-ammonium chloride, diazanium;hexachloroosmium 2-, diammonium hexachloroosmiumdiuide, ammonium hexachloroosmiate iv , premion | |
JRHMYOOMZKAPKT-UHFFFAOYSA-J | |
diazanium;hexachloroosmium(2-) | |
11729867 | |
Hygroscopic | |
Ammonium hexachloroosmate(IV) |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 235-188-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only