Learn More
Ammonium hexafluorotitanate(IV), 99.99%, (trace metal basis), Thermo Scientific Chemicals
£40.90 - £599.99
Chemical Identifiers
CAS | 16962-40-6 |
---|---|
Molecular Formula | H8F6N2Ti |
Molecular Weight (g/mol) | 197.97 |
MDL Number | MFCD00010888 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10432201
|
Thermo Scientific Acros
194350100 |
10 g | Glass bottle |
£40.90
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10194160
|
Thermo Scientific Acros
194350500 |
50 g | Glass bottle |
£136.68
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10677684
|
Thermo Scientific Acros
|
250 g | Glass bottle |
£599.99
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysisChemical Identifiers
16962-40-6 | |
H8F6N2Ti | |
197.97 | |
MFCD00010888 |
Specifications
16962-40-6 | |
Crystalline Powder, Crystals and/or Chunks | |
H8F6N2Ti | |
MFCD00010888 | |
197.97 | |
99.99% | |
Ammonium hexafluorotitanate(IV) |
White | |
99.99% | |
(NH4)2TiF6 | |
Solubility in water: insoluble | |
197.97 | |
Trace Metal Basis |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 241-036-9
- Ammonium hexafluorotitanate(IV)
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only