missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Hydrogen Difluoride, Extra Pure, SLR, Fisher Chemical™
43.91 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 g
2.5 kg
Specifications
Boiling Point | 230°C |
Melting Point | 125°C |
Color | White |
pH | 3.5 |
Physical Form | Solid |
Quantity | 500 g |
Formula Weight | 57.04g/mol |
Packaging | Plastic powder jar |
Vapor Pressure | 1 hPa at 20°C |
Safety and Handling
Product Identifier
- Ammonium hydrogen difluoride
- Danger
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- H301-Toxic if swallowed.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'