Learn More
Ammonium hydrogen fluoride, tech. 95%, Thermo Scientific Chemicals
£31.90 - £83.00
Chemical Identifiers
CAS | 1341-49-7 |
---|---|
Molecular Formula | F2H5N |
Molecular Weight (g/mol) | 57.04 |
MDL Number | MFCD00012018 |
InChI Key | KVBCYCWRDBDGBG-UHFFFAOYSA-N |
Synonym | Ammonium bifluoride |
IUPAC Name | amine dihydrofluoride |
SMILES | N.F.F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11386968
|
Thermo Scientific Alfa Aesar
014660.36 |
500 g |
£31.90
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11396968
|
Thermo Scientific Alfa Aesar
014660.A3 |
2 kg |
£83.00
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ammonium hydrogen fluoride is used as a chemical reagent in analysis and an antiseptic. It is also used in ceramics, electroplating, component of glass etchants as well as food processing equipment disinfectant. It acts as an intermediate in the production of hydrofluoric acid from hexafluorosilicic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1341-49-7 | |
57.04 | |
KVBCYCWRDBDGBG-UHFFFAOYSA-N | |
amine dihydrofluoride |
F2H5N | |
MFCD00012018 | |
Ammonium bifluoride | |
N.F.F |
Specifications
238°C | |
1341-49-7 | |
Flakes | |
MFCD00012018 | |
14,498 | |
Soluble in water. Slightly soluble in alcohol. | |
N.F.F | |
57.04 | |
95% | |
Ammonia-like | |
1.500 g/mL |
120°C to 122°C | |
2 to 3 | |
F2H5N | |
UN1727 | |
Ammonium bifluoride | |
KVBCYCWRDBDGBG-UHFFFAOYSA-N | |
amine dihydrofluoride | |
57.04 | |
Technical | |
Hygroscopic | |
Ammonium hydrogen fluoride |
Safety and Handling
GHS H Statement
H300-H314-H318
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMMONIUM HYDROGENDIFLUORIDE, SOLID
missing translation for 'einecsNumber' : 215-676-4
missing translation for 'rtecsNumber' : BQ9200000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only