missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydrogen sulfate, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11322438
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Ammonium hydrogen sulfate is used as pharmaceutical intermediates and chemical reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7803-63-6
Molecular Formula H5NO4S
Molecular Weight (g/mol) 115.10
MDL Number MFCD00051073
InChI Key BIGPRXCJEDHCLP-UHFFFAOYSA-N
Synonym ammonium bisulfate, ammonium hydrogensulphate, sulfuric acid, monoammonium salt, unii-6218r7mbzb, sulfuric acid, ammonium salt 1:1, azanium hydrogen sulfate, ammonia sulfate, acmc-1bcew, ammonium hydrogen sulphate, nh4hso4
PubChem CID 24655
SMILES [NH4+].OS([O-])(=O)=O

Specifications

Melting Point 146.9°C
Physical Form Lump
Quantity 25 g
UN Number UN2506
Solubility Information Soluble in water (∽115.1mg/ml at 20°C). Insoluble in acetone,alcohol,and pyridine.
IUPAC Name azanium;hydrogen sulfate
Formula Weight 115.1
Percent Purity 99.9%
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Ammonium hydrogen sulfate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'