missing translation for 'onlineSavingsMsg'
Learn More
Ammonium Hydroxide, 30%, BAKER INSTRA-ANALYZED Reagent, J.T. Baker™ Product Code.: 15528654

Ammonium Hydroxide, 30%, BAKER INSTRA-ANALYZED Reagent, J.T. Baker™

Product Code. 15528654
500 mL, Poly container
Click to view available options
Quantity:
500 mL
Packaging:
Poly container
This item is not returnable. View return policy

Product Code. 15528654

Brand: Avantor J.T.Baker 973301

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 1336-21-6
Assay Percent Range 28.0 to 30.0 %
Molecular Formula H5NO
MDL Number MFCD00066650
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Quantity 500 mL
SMILES N.O
IUPAC Name azanium;hydroxide
Molecular Weight (g/mol) 35.05
PubChem CID 14923
ChEBI CHEBI:18219
Formula Weight 35.05
Percent Purity 28.0 to 30.0%
Packaging Poly container
Chemical Name or Material Ammonium Hydroxide, 30%
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium Hydroxide, 30%, BAKER INSTRA-ANALYZED Reagent, J.T. Baker™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.