missing translation for 'onlineSavingsMsg'
Learn More
Ammonium hydroxide, for analysis, 28-30 wt% solution of NH3 in water, Thermo Scientific Chemicals Product Code.: 15294674

Ammonium hydroxide, for analysis, 28-30 wt% solution of NH3 in water, Thermo Scientific Chemicals

Product Code. 15294674
25 L, Plastic Can
Click to view available options
Quantity:
1 L
2.5 L
10 L
25 L
Packaging:
Glass bottle
Plastic Can
Plastic drum
This item is not returnable. View return policy

Product Code. 15294674

Brand: Thermo Scientific Acros 205840250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1336-21-6, 7732-18-5
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 ohShow MoreShow Less
PubChem CID 14923
ChEBI CHEBI:18219
SMILES N.O

Specifications

Boiling Point 38.0°C
Melting Point -57.0°C
Linear Formula NH4OH
Density 0.9100g/mL
Solubility Information Solubility in water: soluble.
Quantity 25 L
IUPAC Name amine hydrate
Formula Weight 35.05
Grade Analytical
Packaging Plastic Can
Specific Gravity 0.91
Chemical Name or Material Ammonium hydroxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467958 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number205840010,205840250,205840025,205840100
Lot NumberA0460066 Certificate TypeCertificate of Analysis Date12/02/2024 Catalog Number205840010,205840250,205840025,205840100
Lot NumberA0459809 Certificate TypeCertificate of Analysis Date02/02/2024 Catalog Number205840010,205840250,205840025,205840100
Lot NumberA0459810 Certificate TypeCertificate of Analysis Date01/02/2024 Catalog Number205840010,205840250,205840025,205840100
Lot NumberA0428274 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog Number205840010,205840250,205840025,205840100

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium hydroxide, for analysis, 28-30 wt% solution of NH3 in water, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.