missing translation for 'onlineSavingsMsg'
Learn More
Ammonium hydroxide, extra pure, 25% solution in water, Thermo Scientific Chemicals Product Code.: 10730751

Ammonium hydroxide, extra pure, 25% solution in water, Thermo Scientific Chemicals

Product Code. 10730751
2.5 L, Glass bottle
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10730751

Brand: Thermo Scientific Acros 255210025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1336-21-6, 7732-18-5
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name amine hydrate
SMILES N.O

Specifications

Color Colorless
Boiling Point 38.0°C
Melting Point -57.0°C
Linear Formula NH4OH
Density 0.9100g/mL
Solubility Information Solubility in water: soluble.
Quantity 2.5 L
Formula Weight 35.05
Percent Purity ≥25% (as NH3) (total base)
Grade Extra Pure
Packaging Glass bottle
Specific Gravity 0.91
Physical Form Liquid
Chemical Name or Material Ammonium hydroxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466151 Certificate TypeCertificate of Analysis Date19/09/2024 Catalog Number255210010,255210100,255210025
Lot NumberA0462439 Certificate TypeCertificate of Analysis Date03/05/2024 Catalog Number255210010,255210100,255210025
Lot NumberA0456243 Certificate TypeCertificate of Analysis Date26/09/2023 Catalog Number255210010,255210100,255210025
Lot NumberA0450538 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number255210010,255210100,255210025
Lot NumberA0446334 Certificate TypeCertificate of Analysis Date14/10/2022 Catalog Number255210010,255210100,255210025
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Ammonium hydroxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium hydroxide, extra pure, 25% solution in water, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.