missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydroxide, extra pure, 25% solution in water, Thermo Scientific Chemicals

Product Code. 11924131
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1336-21-6, 7732-18-5
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name amine hydrate
SMILES N.O

Specifications

Color Colorless
Boiling Point 38.0°C
Melting Point -57.0°C
Linear Formula NH4OH
Density 0.9100g/mL
Solubility Information Solubility in water: soluble.
Quantity 10 L
Formula Weight 35.05
Percent Purity ≥25% (as NH3) (total base)
Grade Extra Pure
Packaging Plastic drum
Specific Gravity 0.91
Physical Form Liquid
Chemical Name or Material Ammonium hydroxide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ammonium hydroxide, 25-30% solution in water
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Ammonium hydroxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.