missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium hydroxide solution, TraceSELECT Ultra, for ultratrace analysis,≥20% in H2O, Fluka, Honeywell
£205.85
Chemical Identifiers
CAS | 7732-18-5, 1336-21-6 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.02 |
MDL Number | MFCD00066650 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | Ammonia aqueous, Ammonia water |
IUPAC Name | water |
SMILES | O |
Description
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
Chemical Identifiers
7732-18-5, 1336-21-6 | |
18.02 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water |
H2O | |
MFCD00066650 | |
Ammonia aqueous, Ammonia water | |
O |
Specifications
24°C | |
≥70.00, ≥20.00 | |
H2O | |
3587154 | |
Ammonia aqueous, Ammonia water | |
O | |
18.02 | |
Poly Bottle |
7732-18-5, 1336-21-6 | |
<90.00, <30.00 | |
MFCD00066650 | |
0.90 g/cm3 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
Trace Analysis | |
Liquid |
Safety and Handling
H314+H335
P261+P264+P271+P280
Danger
Recommended Storage : Room Temperature