missing translation for 'onlineSavingsMsg'
Learn More
Ammonium hydroxide, Thermo Scientific Chemicals Product Code.: 15522233

Ammonium hydroxide, Thermo Scientific Chemicals

Product Code. 15522233
1 L
Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy

Product Code. 15522233

Brand: Thermo Scientific Acros 458680010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 ohShow MoreShow Less
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name amine hydrate
SMILES N.O

Specifications

Linear Formula NH4OH
Density 0.9000g/mL
Quantity 1 L
Formula Weight 35.05
Grade Extra Pure
Specific Gravity 0.9
Chemical Name or Material Ammonium hydroxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0450077 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number458680010,458680025
Lot NumberA0447720 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number458680010,458680025
Lot NumberA0392754 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number458680010,458680025
Lot NumberA0408502 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number458680010,458680025
Lot NumberA0444088 Certificate TypeCertificate of Analysis Date07/10/2022 Catalog Number458680010,458680025
compliance-icons compliance-icons
Product Identifier
  • Ammonium hydroxide, 10% in water
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : ammonium hydroxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium hydroxide, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.