Learn More
Ammonium iodide, 98+%, Thermo Scientific Chemicals
£51.15 - £732.65
Chemical Identifiers
CAS | 12027-06-4 |
---|---|
Molecular Formula | H4IN |
Molecular Weight (g/mol) | 144.94 |
MDL Number | MFCD00011424 |
InChI Key | UKFWSNCTAHXBQN-UHFFFAOYSA-N |
Synonym | ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i |
PubChem CID | 25487 |
ChEBI | CHEBI:63040 |
IUPAC Name | azanium;iodide |
SMILES | [NH4+].[I-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11454134
|
Thermo Scientific Alfa Aesar
A12498.22 |
100 g |
£51.15
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11464134
|
Thermo Scientific Alfa Aesar
A12498.36 |
500 g |
£174.87
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11474134
|
Thermo Scientific Alfa Aesar
A12498.0E |
2500 g |
£732.65
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ammonium iodide is a laboratory reagent and used as a photographic chemicals. It has wide range of applications in chemical industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAmmonium iodide is a laboratory reagent and used as a photographic chemicals. It has wide range of applications in chemical industries.
Solubility
Soluble in water (1770 mg/ml at 25°C), methanol (400 mg/ml), glycerol (667 mg/ml), alcohol (270 mg/ml), and ethanol.
Notes
Store at 4°C. Hande under dry protective gas.
Chemical Identifiers
12027-06-4 | |
144.94 | |
UKFWSNCTAHXBQN-UHFFFAOYSA-N | |
25487 | |
azanium;iodide |
H4IN | |
MFCD00011424 | |
ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i | |
CHEBI:63040 | |
[NH4+].[I-] |
Specifications
551°C (sublimation) | |
H4IN | |
14,529 | |
Soluble in water (1770mg/ml at 25°C),methanol (400mg/ml),glycerol (667mg/ml),alcohol (270mg/ml),and ethanol. | |
[NH4+].[I-] | |
144.94 | |
CHEBI:63040 | |
≥98% | |
1.7031 | |
2.514 |
12027-06-4 | |
MFCD00011424 | |
ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i | |
UKFWSNCTAHXBQN-UHFFFAOYSA-N | |
azanium;iodide | |
25487 | |
144.94 | |
Odorless | |
Air and light sensitive; Hygroscopic | |
Ammonium iodide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 234-717-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only