missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Iodide, ACS Reagent, Honeywell Fluka™

ACS Reagent,≥99%

£40.30 - £145.76

Chemical Identifiers

CAS 12027-06-4
Molecular Formula H4IN
Molecular Weight (g/mol) 144.94
MDL Number MFCD00011424
InChI Key UKFWSNCTAHXBQN-UHFFFAOYSA-N
Synonym ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i
PubChem CID 25487
ChEBI CHEBI:63040
IUPAC Name ammonium iodide
SMILES [NH4+].[I-]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15602170
View Documents Promotion Details
Honeywell-Fluka
221937-100G
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15612170
View Documents Promotion Details
Honeywell-Fluka
221937-500G
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

12027-06-4
144.94
UKFWSNCTAHXBQN-UHFFFAOYSA-N
25487
ammonium iodide
H4IN
MFCD00011424
ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i
CHEBI:63040
[NH4+].[I-]

Specifications

12027-06-4
NH4I
NONH for all modes of transport
UKFWSNCTAHXBQN-UHFFFAOYSA-N
ammonium iodide
25487
144.94g/mol
ACS Reagent
1mmHg (210.9°C)
H4IN
MFCD00011424
ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i
[NH4+].[I-]
144.94
CHEBI:63040
≥99%
≤0.05%
Ammonium Iodide
Videos
Safety and Handling

Safety and Handling

P261-P305 + P351 + P338

H315-H319-H335

missing translation for 'einecsNumber' : 234-717-7

SDS
Documents

Documents

Product Certifications