missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Iodide, puriss., Honeywell Fluka™
Click to view available options
Quantity:
100 g
1 kg
50 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 1 kg |
Linear Formula | NH4I |
UN Number | NONH for all modes of transport |
IUPAC Name | azanium;iodide |
Formula Weight | 144.94g/mol |
Percent Purity | ≥99% |
Grade | puriss. |
Residue after Ignition | ≤0.1% (as SO4) |
Vapor Pressure | 1mmHg (210.9°C) |
Chemical Name or Material | Ammonium Iodide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'