missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Iodide, puriss. p.a. ACS Reagent, ≥99.0% (AT), Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Color | White |
pH | 4.5 to 6.5 (at 20°C) |
Quantity | 100 g |
Linear Formula | NH4I |
UN Number | NONH for all modes of transport |
Solubility Information | Soluble in water, 50mg/mL |
Formula Weight | 144.94g/mol |
Percent Purity | ≥99.0% (AT) |
Grade | ACS/puriss. p.a. |
Packaging | Glass Bottle |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'