missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iron(II) sulfate hexahydrate, 99+%, for analysis, Thermo Scientific Chemicals

£24.52 - £629.13

Chemical Identifiers

CAS 7783-85-9
Molecular Formula FeH20N2O14S2
Molecular Weight (g/mol) 392.13
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
Synonym Ferrous ammonium sulfate, Mohr's salt
IUPAC Name λ²-iron(2+) diammonium hexahydrate disulfate
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10762241
View Documents
Thermo Scientific Acros
201370250
25 g Plastic bottle
£24.52
25g
Please to purchase this item. Need a web account? Register with us today!
10532991
View Documents
Thermo Scientific Acros
201375000
500 g Plastic bottle
£37.19
500g
Please to purchase this item. Need a web account? Register with us today!
10052750
View Documents Promotion Details
Thermo Scientific Acros
201370010
1 kg Plastic bottle
£55.38
1kg
Please to purchase this item. Need a web account? Register with us today!
10553124
View Documents
Thermo Scientific Acros
201370050
5 kg Plastic bottle
£187.94
5kg
Please to purchase this item. Need a web account? Register with us today!
10444985
View Documents
Thermo Scientific Acros
201370251
25 kg Fibre drum
£629.13
25kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
Specifications

Chemical Identifiers

7783-85-9
392.13
MQLVWQSVRZVNIP-UHFFFAOYSA-L
λ²-iron(2+) diammonium hexahydrate disulfate
FeH20N2O14S2
MFCD00150530
Ferrous ammonium sulfate, Mohr's salt
[NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

100.0°C
Green
99+%
(NH4)2Fe(SO4)2·6H2O
15, 518
Solubility in water: 269g/L (20°C). Other solubilities: practically insoluble in alcohol
[NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
392.13
≥99%
Ammonium iron(II) sulfate hexahydrate
7783-85-9
Crystalline Powder and/or Crystals
FeH20N2O14S2
MFCD00150530
Ferrous ammonium sulfate, Mohr's salt
MQLVWQSVRZVNIP-UHFFFAOYSA-L
λ²-iron(2+) diammonium hexahydrate disulfate
392.14
Analytical
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Recommended Storage : Due to inherent oxidation, ferric ion (Fe3+) content may increase on storage

Product Identifier
  • Ammonium iron (II) sulfate hexahydrate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only