missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iron(III) citrate, 16.5 to 18.5% Fe, Thermo Scientific Chemicals

Product Code. 11985061
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185-57-5
Molecular Formula C6H11FeNO7
Molecular Weight (g/mol) 265
MDL Number MFCD00013099
InChI Key FRHBOQMZUOWXQL-UHFFFAOYSA-N
Synonym ammonium ferric citrate, ferri seltz, ammonii ferri citras, ferric ammonium citrate, iron ammonium citrate, ammonium iron 3+ citrate, ammonium iron iii citrate, iron ammonium citrate, green, ferri-ammoniumcitrat, braunes, ferriseltz
PubChem CID 118984355
IUPAC Name azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+)
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3]

Specifications

Color Brown to Brown-Yellow
Physical Form Powder
Quantity 10 kg
Merck Index 15, 4044
Solubility Information Solubility in water: 1200g/L (20 c). Other solubilities: practically insoluble in alcohol
Formula Weight 265
Percent Purity 16.5 to 18.5% (Fe)
Infrared Spectrum Authentic
Packaging Plastic drum
Chemical Name or Material Ammonium iron(III) citrate, 16.5 to 18.5% Fe

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'