missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iron(III) citrate, 16.5 to 18.5% Fe

Product Code. p-4509959 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
250g
This item is not returnable. View return policy

Product Code. 10762505

Brand: Thermo Scientific Acros 211842500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1185-57-5 | C6H11FeNO7 | 265 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185-57-5
Molecular Formula C6H11FeNO7
Molecular Weight (g/mol) 265
MDL Number MFCD00013099
InChI Key FRHBOQMZUOWXQL-UHFFFAOYSA-N
Synonym ammonium ferric citrate, ferri seltz, ammonii ferri citras, ferric ammonium citrate, iron ammonium citrate, ammonium iron 3+ citrate, ammonium iron iii citrate, iron ammonium citrate, green, ferri-ammoniumcitrat, braunes, ferriseltz
PubChem CID 118984355
IUPAC Name azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+)
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3]

Specifications

Color Brown to Brown-Yellow
Physical Form Powder
Quantity 250 g
Merck Index 15, 4044
Solubility Information Solubility in water: 1200g/L (20 c). Other solubilities: practically insoluble in alcohol
Formula Weight 265
Percent Purity 16.5 to 18.5% (Fe)
Infrared Spectrum Authentic
Packaging Plastic bottle
Chemical Name or Material Ammonium iron(III) citrate, 16.5 to 18.5% Fe

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.