missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Iron(Iii) Citrate, Honeywell™
Click to view available options
Quantity:
50 g
500 g
1 kg
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Green |
Physical Form | Powder |
Quantity | 1 kg |
Linear Formula | C6H8O7 ·xFe3+ · yNH3 |
Grade | purum p.a. |
Concentration or Composition (by Analyte or Components) | 14.5 to 16% Fe basis (RT) |
Chemical Name or Material | Ammonium Iron(Iii) Citrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'