missing translation for 'onlineSavingsMsg'
Learn More
Ammonium Iron(Iii) Citrate, Honeywell™ Product Code.: 15630890

Ammonium Iron(Iii) Citrate, Honeywell™

Product Code. 15630890 Shop All Honeywell Research Chemicals Products
50 g
Click to view available options
Quantity:
50 g
500 g
1 kg
This item is not returnable. View return policy

Product Code. 15630890

Brand: Honeywell Research Chemicals 0971350G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Purum p.a., 14.5-16% Fe basis (RT), powder, green

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185-57-5
Molecular Formula C6H11FeNO7+3
Molecular Weight (g/mol) 264.999
InChI Key FRHBOQMZUOWXQL-UHFFFAOYSA-N
Synonym ammonium ferric citrate, ferri seltz, ammonii ferri citras, ferric ammonium citrate, iron ammonium citrate, ammonium iron 3+ citrate, ammonium iron iii citrate, iron ammonium citrate, green, ferri-ammoniumcitrat, braunes, ferriseltz
PubChem CID 118984355
IUPAC Name azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+)
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3]

Specifications

Color Green
Physical Form Powder
Quantity 50 g
Linear Formula C6H8O7 ·xFe3+ · yNH3
Grade purum p.a.
Concentration or Composition (by Analyte or Components) 14.5 to 16% Fe basis (RT)
Chemical Name or Material Ammonium Iron(Iii) Citrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium Iron(Iii) Citrate, Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.