missing translation for 'onlineSavingsMsg'
Learn More
Ammonium iron(III) oxalate hydrate, Thermo Scientific Chemicals Product Code.: 11389767

Ammonium iron(III) oxalate hydrate, Thermo Scientific Chemicals

Product Code. 11389767
50 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11389767

Brand: Thermo Scientific Alfa Aesar 042112.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

In photography, blueprints

In photography, blueprints

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13268-42-3
Molecular Formula C6H18FeN3O15
Molecular Weight (g/mol) 428.06
MDL Number MFCD00150137
InChI Key ARJSTCBPHIPXIJ-UHFFFAOYSA-K
Synonym Ferric ammonium oxalate; Iron(III) ammonium oxalate
IUPAC Name triammonium hexaoxo-1,4,6,9,10,13-hexaoxa-5-ferraspiro[4.4⁵.4⁵]tridecane-5,5,5-tris(ylium) trihydrate
SMILES [NH4+].[NH4+].[NH4+].O.O.O.O=C1O[Fe+3]234(OC1=O)OC(=O)C(=O)O2.O=C(O3)C(=O)O4

Specifications

Melting Point (decomposition)
Quantity 50 g
UN Number UN3288
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,517
Solubility Information Very soluble in water; Insoluble in alcohol
Formula Weight 374.02 (Anhydrous)
Physical Form Powder
Chemical Name or Material Ammonium iron(III) oxalate hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberH18Q11 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number042112.18,042112.30,042120.09
Lot NumberH23K14 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number042112.18,042112.30,042120.09
Lot Number2628 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number042112.18,042112.30,042120.09
Lot NumberA08M17 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number042112.18,042112.30,042120.09
Lot NumberI06Z044 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number042112.18,042112.30,042120.09
compliance-icons
Product Identifier
  • Ammonium iron(III) oxalate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P363-Wash contaminated clothing before reuse.
  • P321-Specific treatment (see on this label).
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium iron(III) oxalate hydrate, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.