missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium iron(III) sulfate dodecahydrate, 99+%, for analysis, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Boiling Point | 230.0°C |
Melting Point | 39.0°C to 41.0°C |
Color | Gray-Purple |
Physical Form | Adhering Crystalline Powder or Crystals |
Quantity | 500 g |
Assay Percent Range | 99% min. (Iodometry) |
Linear Formula | (NH4)Fe(SO4)2·12H2O |
Merck Index | 15, 515 |
Solubility Information | Solubility in water: 1240g/L water (25°C). Other solubilities: practically insoluble in alcohol |
Formula Weight | 482.19 |
Show More |
Safety and Handling
Product Identifier
- Ammonium iron(III) sulfate dodecahydrate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Iron(III) ammonium sulfate dodecahydrate
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'