missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium iron(III) sulfate dodecahydrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., 99.0-102.0%, crystalline, slightly violet, Honeywell Fluka™
Click to view available options
Quantity:
500 g
2.5 kg
50 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 39°C to 41°C (literature) |
Color | Violet |
CAS Max % | 100.0000% |
Physical Form | Crystalline |
Quantity | 50 kg |
Assay Percent Range | 99.0 to 102.0% |
Linear Formula | Na4P2O7 · 10H2O |
MDL Number | MFCD00149200 |
UN Number | NONH for all modes of transport |
Solubility Information | In accordance with water, 10g/dL |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'